Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromothymol blue 0.04% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical
Synonyms: 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide3′,3″-Dibromothymolsulfonphthalein3′,3″-Dibromothymolsulphonphthalein

R1375000-1A R1375000-500A R1375000-120A
RC137516EA 61.22 USD InStock
RC137516 RC137532 RC13754
Bromothymol blue 0.04% (w/v) in aqueous solution indicator
Bromothymol blue


Formula: C₂₇H₂₈Br₂O₅S
MW: 624.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005872
CAS Number: 76-59-5
EINECS: 200-971-2
Order Now
Specification Test Results
Appearance Clear, green, odorless
pH at 25 °C (traceable to NIST SRM 185 & 186) 6.8 ± 0.1 pH at 25.0 °C
Suitability for Use Yellow (pH 7.6)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty