Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromothymol blue 0.1% (w/v) in 50% (v/v) alcohol USP Test Solution (TS)

Supplier: Ricca Chemical
Danger
Synonyms: 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide3′,3″-Dibromothymolsulfonphthalein3′,3″-Dibromothymolsulphonphthalein

R1378000-500A R1378000-120A
RC1378-4EA 118.72 USD InStock
RC1378-4 RC1378-16
Bromothymol blue 0.1% (w/v) in 50% (v/v) alcohol USP Test Solution (TS)
Bromothymol blue


Formula: C₂₇H₂₈Br₂O₅S
MW: 624.39 g/mol
Boiling Pt: ~ 82 °C
Density: 0.9000 g/cm³
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00005872
CAS Number: 76-59-5
UN: 1987
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear, reddish orange, ethanol odor
Suitability for Use Yellow (pH 7.6)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty