Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromothymol blue-phenol red mixed indicator

Supplier: Ricca Chemical

R1385000-120A R1385000-500A
RC1385-16EA 74.05 USD InStock
RC1385-16 RC1385-4
Bromothymol blue-phenol red mixed indicator
Bromothymol blue-phenol red


Order Now
Specification Test Results
Appearance Dark green to purplish red, odorless
pH at 25 °C (traceable to NIST SRM 186 & 191) 7.50 ± 0.05 pH at 25.0 °C
Suitability for Use Yellow (pH 7.5)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty