Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buffers, Reference standards, Acidic to Neutral, Ricca Chemical

Supplier: RICCA Chemical Company

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

R1498000-1A R1550000-4A R1489000-20F R1490000-4A 1485-32 R1495000-1A R1540000-4A R1492000-4A R1490000-500A R1498000-500A R1495000-4A R1498000-4A R1492000-1A R1505000-4A R1550000-10F R1565000-1A R1565000-4A R1493000-4A R1495000-20PK R1550000-4F R1540000-500A R1540000-20F R1495000-10F R1540000-1A 1485-16 R1492000-10F R1550000-1A R1510000-10F R1505000-1A R1510000-4A R1498000-20F R1489000-1A 1563-16 R1500000-4F R1489000-500A R1540000-10F R1500000-500A R1493000-500A R1565000-20F R1565000-500A 1485-1 R1500000-4A R1492000-20PK R1492000-500A R1493000-20F R1492000-20F R1493000-10F R1550000-500A R1493000-20PK R1490000-1A R1505000-10F R1500000-20F R1550000-20F R1489000-4A R1495000-20F R1493000-1A R1505000-500A R1495000-500A R1510000-1A R1500000-10F R1510000-500A R1500000-1A R1505000-20F
RC1495-16CS 1389.69 USD InStock
RC1495-16 RC1485-1 RCR1540000-20F RC1489-16 RC1489-1 RC1540-32 RCR1565000-20F RC155016 RC1510-1 RC15505 RC1492-32 RC15501 RC1493-5 RC1490-32 RC1495-1 RC15001 RC1493-1 RC1505-32 RC1563-16 RC1495-5 RCR1565000-1A RCR1489000-20F RC155025 RCR149500020PK RCR1498000-20F RCR1510000-10F RC154016 RC1485-16 RC1540-1 RCR1505000-10F RC150016 RC1510-16 RC1493-32 RCR149300020PK RCR1495000-10F RC1500-1CT RCR1493000-10F RC1489-32 RC1493-16 RCR1540000-10F RC155032 RC1485-32 RCR1565000-4A RC1550-1CT RC150025 RC1498-32 RC15005 RC1498-1 RC1505-16 RC1492-5 RC149216 RC149016 RC1492-1 RCR1565000500A RC149816 RC150032 RCR1505000-20F RCR149200020PK RC1490-1 RC1510-32 RC1505-1 RC1495-32 RCR1492000-10F
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Buffers pH Calibration Buffers
Colorless pH buffer solutions are certified to ±0.01 pH at 25°C

  • Range of packaging is available

Each 500 ml plastic bottle is labeled with the certified pH value, the NIST SRM number and a pH/temperature conversion.

Certifications: Standardized against solutions whose certification is traceable to the National Institute of Standards and Technology. Meet USP requirements.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty