Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride 10 g/L in mixed alcohol solvent for determination of salts in crude oil

Supplier: Ricca Chemical
Danger
Synonyms: Chloro calcium

R1718000-1C
RC1718-32EA 128.78 USD InStock
RC1718-32
Calcium chloride 10 g/L in mixed alcohol solvent for determination of salts in crude oil
Calcium chloride


Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: ~ 118 °C
Density: 0.8700 g/cm³
Flash Pt: ~ 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
UN: 1987
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear, colorless, Butanol odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty