Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride 0.100 M (M/10) in aqueous solution

Supplier: Ricca Chemical
Warning
Synonyms: Chloro calcium

R1760000-1A R1760000-500A
RC176016EA 59.83 USD InStock
RC176016 RC176032
Calcium chloride 0.100 M (M/10) in aqueous solution
Calcium chloride


Formula: CaCl₂
MW: 110.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
Order Now
Specification Test Results
Appearance Clear, colorless lquid
Assay at 20 °C (traceable to NIST SRM 915) 0.1000 ± 0.0001 M at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty