Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride 7.5% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Danger
Synonyms: Chloro calcium

R1783000-1A R1783000-500A
RC1783-16EA 97.27 USD InStock
RC1783-16 RC1783-32
Calcium chloride 7.5% (w/v) in aqueous solution USP Test Solution (TS)
Calcium chloride


Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0456 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 7.50 ± 0.01 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty