Would you like to log in again?
Calcium IC Standard, 1 ml = 0.100 mg Ca, 100 ppm Ca CaCO₃in 3% HNO₃, Ricca Chemical Company
Supplier: RICCA Chemical Company
R1795000-100A
RC1795-100EA 49.71 USD InStock
RC1795-100
Calcium IC Standard, 1 ml = 0.100 mg Ca, 100 ppm Ca CaCO₃in 3% HNO₃, Ricca Chemical Company
Standards Cationic Standards Calcium Standards
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty