Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium sulfate solution R, saturated

Supplier: Ricca Chemical

R1823000-500A R1823000-120A
RC1823-16EA 252.74 USD InStock
RC1823-16 RC1823-4
Calcium sulfate solution R, saturated
Calcium sulphate


Formula: CaO₄S
MW: 136.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Calcium Present
Sulfate Present

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty