Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calmagite 0.1% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical

R1830000-500A R1830000-120A
RC1830-4EA 42.43 USD InStock
RC1830-4 RC183016
Calmagite 0.1% (w/v) in aqueous solution indicator
Calmagite
APHA, for water hardness.


Formula: C₁₇H₁₄N₂O₅S
MW: 358.37
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011656
CAS Number: 3147-14-6
Order Now
Specification Test Results
Appearance Dark red, characteristic Calmagite odor
Suitability for Use Wine Red (Excess Ca/Mg) - Pure Blue (Excess EDTA)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty