Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carmine reagent for determination of boron

Supplier: Ricca Chemical
Danger
Synonyms: Alum lake of Carminic AcidCarmine, calcium/aluminum lakeCI 75470Natural Red 4Carminic acidCochinealFD and C Carmine

R1850000-500C1 R1850000-120C R1850000-1C1
RC1850-16EA 177.33 USD InStock
RC1850-16 RC1850-32 RC1850-4
Carmine reagent for determination of boron
Carmine
Carmine Reagent, for Boron determination

  • APHA Standard Methods Reagents
  • Shelf Life 12 months


Formula: C₂₂H₂₀O₁₃
MW: 492.389 g/mol
Boiling Pt: ~ 338 °C
Density: 1.8000 g/cm³
Storage Temperature: Ambient
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, red, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty