Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorine water saturated ∼0.3% (w/v)

Supplier: Ricca Chemical
Warning

1980-32 1980-16
RC1980-32EA 132.7 USD InStock
RC1980-32 RC1980-16
Chlorine water saturated ∼0.3% (w/v)
Chlorine water


Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Storage Temperature: Ambient
CAS Number: 7782-50-5
UN: 1760
ADR: 8,III
Order Now
Specification Test Results
Appearance Yellow-green liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty