Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorophenol red 0.04% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical
Synonyms: 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

R2000000-500A R2000000-120A
RC200016EA 60.28 USD InStock
RC200016 RC20004
Chlorophenol red 0.04% (w/v) in aqueous solution indicator
Chlorophenol red


Formula: C₁₉H₁₂Cl₂O₅S
MW: 423.27 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005877
CAS Number: 4430-20-0
Order Now
Specification Test Results
Appearance Deep red, odorless
Suitability for Use Yellow (pH 6.4)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty