Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chromicsulfuric acid solution for cleaning of laboratory glassware

Supplier: Ricca Chemical
Danger

R2150000-1C1 R2150000-2.5D
RC215032EA 120.61 USD InStock
RC215032 RC21505PT
Chromicsulfuric acid solution for cleaning of laboratory glassware
Chromicsulfuric acid solution
For cleaning glassware.


Boiling Pt: ~ 300 °C
Density: 1.8000 g/cm³
Storage Temperature: Ambient
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Appearance Dark brown, viscous liquid, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty