Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt(II) chloride 1.2% (w/v) in hydrochloric acid 10%

Supplier: Ricca Chemical
Danger

R2180000-500A
RC218016EA 80.8 USD InStock
RC218016
Cobalt(II) chloride 1.2% (w/v) in hydrochloric acid 10%
Cobalt(II) chloride solutions


Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0300 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
UN: 3264
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, dark pink, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty