Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt(II) chloride 2% (w/v) in diluted hydrochloric acid USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R2208000-500A
RC2208-16EA 255.31 USD InStock
RC2208-16
Cobalt(II) chloride 2% (w/v) in diluted hydrochloric acid USP Test Solution (TS)
Cobalt(II) chloride solutions


Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
Order Now
Specification Test Results
Appearance Clear, light red, odorless
Assay at 20 °C (traceable to NIST SRM 915) 2.00 ± 0.02 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty