Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Color reagent for nitrate determination

Supplier: Ricca Chemical
Danger

R2233000-500C R2233000-120C
RC2233-4EA 70.47 USD InStock
RC2233-4 RC2233-16
Color reagent for nitrate determination
Color reagent


Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0500 g/cm³
Storage Temperature: Refrigerator
UN: 3264
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, tan-brown, odorless
Suitability for Use Red color with Nitrite

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty