Would you like to log in again?
Conductivity/TDS Standard, 20 mS/cm (20000 µmho/cm) at 25 °C, 11000 ppm as NaCl, Ricca Chemical Company
Supplier: RICCA Chemical Company
R2247000-4A R2247000-500A R2247000-1A
RC224716EA 51.3 USD InStock
RC224716 RC22471 RC224732
Conductivity/TDS Standard, 20 mS/cm (20000 µmho/cm) at 25 °C, 11000 ppm as NaCl, Ricca Chemical Company
Standards Conductivity Standards
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty