Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Congo red USP Test Solution (TS)

Supplier: Ricca Chemical
Danger
Synonyms: Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulfonate)Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate)Direct Red 28Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulfonate)Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate)CI 22120

R2253000-120A R2253000-500A
RC2253-16EA 135.02 USD InStock
RC2253-16 RC2253-4
Congo red USP Test Solution (TS)
Congo Red


Formula: C₃₂H₂₂N₆Na₂O₆S₂
MW: 696.67 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 0.9600 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004028
CAS Number: 573-58-0
Order Now
Specification Test Results
Appearance Clear, red, acohol odor
Suitability for Use Red (pH 5.0) - Blue (pH 3.0)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty