Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

m-Cresol purple sodium salt 0.04% (w/v) in aqueous solution pH-indicator

Supplier: Ricca Chemical
Synonyms: 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[d][1,2]oxathiole 2,2-dioxide, sodium saltm-Cresolsulfonphthalein sodium saltm-Cresolsulphonphthalein sodium salt

R2400000-120A R2400000-500A
RC24004EA 36.37 USD InStock
RC24004 RC240016
m-Cresol purple sodium salt 0.04% (w/v) in aqueous solution pH-indicator
m-Cresol purple sodium salt


Formula: C₂₁H₁₇NaO₅S
MW: 404.42 g/mol
MDL Number: MFCD00010177
CAS Number: 62625-31-4
EINECS: 204-087-8
Order Now
Specification Test Results
Appearance Yellow, odorless
Suitability for Use Purple (pH 9.0) - Yellow (pH 2.8 to pH 7.4) - Red (pH 1.2)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty