Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) acetate USP Test Solution (TS), strong

Supplier: Ricca Chemical
Danger

R2488000-500A
RC2488-16EA 249.27 USD InStock
RC2488-16
Copper(II) acetate USP Test Solution (TS), strong
Copper(II) acetate solutions


Formula: (H₃CCOO)₂Cu·H₂O
MW: 199.65 g/mol
Boiling Pt: ~ 100 °C
Density: 1.0677 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149570
CAS Number: 6046-93-1
Order Now
Specification Test Results
Appearance Clear, blue. acetate odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty