Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cupric citrate USP Test Solution (TS), alkaline

Supplier: Ricca Chemical
Danger
Synonyms: Copper(II) citrateCopper citrateCuprocitrol

R2493000-500A R2493000-120A
RC2493-16EA 341.19 USD InStock
RC2493-16 RC2493-4
Cupric citrate USP Test Solution (TS), alkaline
Cupric citrate


Formula: C₆H₄Cu₂O₇
MW: 315.18 g/mol
Boiling Pt: ~ 100 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
CAS Number: 866-82-0
Order Now
Specification Test Results
Appearance Clear, blue, odorless
Suitability for Use Suitable for reducing sugar determination

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty