Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper (II) ethylenediamine complex 1 Molar

Supplier: Ricca Chemical
Danger
Synonyms: CupriethylendiamineBis(ethylenediamine)copper dihydroxideCopper(II)-ethylenediame complex

2499-32 2499-1
RC2499-32EA 182.23 USD InStock
RC2499-32 RC2499-1
Copper (II) ethylenediamine complex 1 Molar
Copper (II) ethylenediamine


Formula: Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MW: 217.76 g/mol
Density: 1.1000
MDL Number: MFCD00274628
CAS Number: 13426-91-0
UN: 1761
ADR: 8 (6.1),III
Order Now
Specification Test Results
Appearance blue-purple liquid
Assay (vs. Sodium Thiosulfate/Starch) 0.98-1.02 Molar
Ethylenediamine / Cupric Ratio 1.96-2.04

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty