Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Dimethylamino)benzaldehyde solution indicator for hydrazine

Supplier: Ricca Chemical
Danger
Synonyms: Ehrlich's Reagent4-(N,N-Dimethylamino)benzaldehydep-Dimethylamino-Benzaldehydep-(Dimethylamino)benzaldehyde

R2555000-500C
RC255516EA 162.74 USD InStock
RC255516
4-(Dimethylamino)benzaldehyde solution indicator for hydrazine
4-(Dimethylamino)benzaldehyde


Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: ~ 71 °C
Density: 0.9000 g/cm³
Flash Pt: ~ 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
UN: 2924
ADR: 3 (8),II
https://us-legacy.vwr.com/stibo/web/std.lang.all/62/54/746254.jpg
Order Now
Specification Test Results
Appearance Clear, light greenish-yellow, Methanol/Hydrochloric odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty