Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-(4-(Dimethylamino)benzylidene)rhodanine 0.02% (w/v) in acetone indicator for argentimetric titrations

Supplier: Ricca Chemical
Danger
Synonyms: 5-(4-Dimethylaminobenzylidene)-2-thioxo-4-thiazolidinonep-Dimethylaminobenzalrhodanine5-(4-(Dimethylamino)benzylidene)-2-mercaptothiazol-4(5H)-onep-(Dimethylamino)benzal-5-rhodanine4-Dimethylaminobenzalrhodanine(5E)-5-[4-(Dimethylamino)benzylidene]-2-mercapto-1,3-thiazol-4(5H)-one

R2560000-500C R2560000-120C
RC2560-16EA 183.93 USD InStock
RC2560-16 RC25604
5-(4-(Dimethylamino)benzylidene)rhodanine 0.02% (w/v) in acetone indicator for argentimetric titrations
5-(4-(Dimethylamino)benzylidene)rhodanine


Formula: C₁₂H₁₂N₂OS₂
MW: 264.37 g/mol
Boiling Pt: 56.05 °C (1013.25 hPa)
Melting Pt: –94.7 °C
Density: 0.7900 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00064857
CAS Number: 536-17-4
UN: 1090
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, light yellow, acetone odor
Suitability for Use Yellow in Absence of Silver Ions to Salmon for an excess of Silver ions

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty