Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl glyoxime solution 1% (w/v) alcoholic solution USP Reference Solution (RS) for nickel determination

Supplier: Ricca Chemical
Danger
Synonyms: Diacetyl dioxime2,3-Butanedione dioximeDiacetyl glyoxime

R2570000-500A R2570000-120A
RC2570-16EA 113.92 USD InStock
RC2570-16 RC2570-4
Dimethyl glyoxime solution 1% (w/v) alcoholic solution USP Reference Solution (RS) for nickel determination
Dimethyl glyoxime


Formula: CH₃C(=NOH)C(=NOH)CH₃
MW: 116.12 g/mol
Boiling Pt: ~ 77 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 15.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00002117
CAS Number: 95-45-4
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, colorless, alcohol odor
Suitability for Use Red precipitate with Nickel

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty