Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,5-Diphenylcarbazide 0.5% (w/v) in acetone

Supplier: Ricca Chemical
Danger
Synonyms: sym. Diphenylcarbazidesym-DiphenylcarbazideN'',N'''-Diphenylcarbonohydrazide1,5-DiphenylcarbohydrazideDiphenylcarbazide1,5-Diphenylcarbonohydrazide

R2590000-500C R2590000-120C
RC2590-16EA 255.64 USD InStock
RC2590-16 RC2590-4
1,5-Diphenylcarbazide 0.5% (w/v) in acetone
1,5-Diphenylcarbazide


Formula: C₁₃H₁₄N₄O
MW: 242.28 g/mol
Boiling Pt: 56.05 °C (1013.25 hPa)
Melting Pt: –94.7 °C
Density: 0.7900 g/cm³
Flash Pt: –20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003013
CAS Number: 140-22-7
UN: 1090
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/46/13/2374613.jpg
Order Now
Specification Test Results
Appearance Clear, colorless to not more than slight yellow-no pink, strong acetone odor
Suitability for Use Red in presence of Chromium

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty