Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acid alcohol 5% HCl (v/v) in 95% denatured alcohol, strong

Supplier: Ricca Chemical
Danger

R0260000-4A R0260000-1A R0260000-500A
RC2601EA 225.61 USD InStock
RC2601 RC26032 RC26016
Acid alcohol 5% HCl (v/v) in 95% denatured alcohol, strong
Acid alcohol


Boiling Pt: ~ 82 °C
Density: 0.8088 g/cm³
Flash Pt: ~ 23.5 °C
Storage Temperature: Ambient
UN: 2924
ADR: 3 (8),II
Order Now
Specification Test Results
Appearance Clear, colorless, strong alcohol odor
Assay at 20 °C (traceable to NIST SRM 84) 5.0 ± 0.1 % (v/v) at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty