Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,5-Diphenylcarbazone solution 1% (w/v) alcoholic solution USP Test Solution (TS)

Supplier: Ricca Chemical
Danger
Synonyms: sym-DiphenylcarbazonePhenylazoformic acid 2-phenylhydrazidPhenylazoameisensäure-2-phenylhydrazidPhenylazoformic acid 2-phenylhydrazide

R2605000-500C R2605000-120C
RC2605-4EA 73.17 USD InStock
RC2605-4 RC2605-16
1,5-Diphenylcarbazone solution 1% (w/v) alcoholic solution USP Test Solution (TS)
1,5-Diphenylcarbazone


Formula: C₁₃H₁₂N₄O
MW: 240.26 g/mol
Boiling Pt: ~ 77 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 15.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003024
CAS Number: 538-62-5
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Appearance Red, alcohol odor
Suitability for Use Colorless (excess Chloride) - Purple (excess Mercury)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty