Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenylcarbazone-bromophenol blue mixed indicator, for chloride determination

Supplier: Ricca Chemical
Danger

R2610000-120C R2610000-500C
RC26104EA 102.17 USD InStock
RC26104 RC261016
Diphenylcarbazone-bromophenol blue mixed indicator, for chloride determination
Diphenylcarbazone-bromophenol blue


Boiling Pt: ~ 77 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 15.5 °C
Storage Temperature: Refrigerator
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Appearance Deep red under light
Suitability for Use Yellow (excess Chloride & proper pH) - Purple (excess Mercury)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty