Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenylcarbazone-xylene cyanol mixed indicator, for low level chloride determination, (not acidifier)

Supplier: Ricca Chemical
Danger

R2620000-120C R2620000-500C
RC26204EA 112.79 USD InStock
RC26204 RC262016
Diphenylcarbazone-xylene cyanol mixed indicator, for low level chloride determination, (not acidifier)
Diphenylcarbazone-xylene cyanol


Boiling Pt: ~ 77 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 15.5 °C
Storage Temperature: Refrigerator
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Appearance Purplish-blue to bluish green, alcohol odor
Suitability for Use Light Green (pH 3.8

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty