Would you like to log in again?
DPD solution indicator, for residual chlorine analysis
Supplier: Ricca Chemical
R2655000-500C R2655000-1C
RC265532EA 204.7 USD InStock
RC265532 RC265516
DPD solution indicator, for residual chlorine analysis
DPD solution
Indicator. APHA, for chlorine.
Order Now
Specification Test Results
Appearance Clear, tan (preferred) to very light pink
Suitability for Use Pink to red color in the presence of Chlorine
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty