Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA disodium salt 10% (w/v) in aqueous solution

Supplier: Ricca Chemical
Synonyms: Ethylenediaminetetra-acetic acid disodium salt dihydrateEdetate disodium dihydrateKestranal® 2S 2 H2OEthylenedinitrilotetra-acetic acid disodium salt dihydrateTitriplex IIIEdetic acid disodium salt dihydrateKestranal® 2S dihydrateDisodium dihydrogen ethylenediaminetetraacetate dihydrate

R2670000-500A R2670000-120A
RC267016EA 87.86 USD InStock
RC267016 RC26704
EDTA disodium salt 10% (w/v) in aqueous solution
EDTA disodium salt


Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0500 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
Order Now
Specification Test Results
Appearance Colorless to slight yellow liquid
Assay at 20 °C (traceable to NIST SRM 915) 10.0 ± 0.4 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty