Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ehrlich aldehyde reagent, Wallace-diamond formulation for urobilinogen determination

Supplier: Ricca Chemical
Danger

R2780000-120C R2780000-500C
RC278016EA 88.38 USD InStock
RC278016 RC27804
Ehrlich aldehyde reagent, Wallace-diamond formulation for urobilinogen determination
Ehrlich aldehyde reagent, Wallace-diamond formulation


Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0500 g/cm³
Storage Temperature: Ambient
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, greenish-yellow, slightly pungent to pungent odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty