Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydrogen carbonate concentrate 0.5 M, 100X eluent

Supplier: Ricca Chemical
Synonyms: Sodium bicarbonate

R2805000-50A
RC2805-50AEA 178.63 USD InStock
RC2805-50A
Sodium hydrogen carbonate concentrate 0.5 M, 100X eluent
Sodium hydrogen carbonate
Sodium carbonate eluent


Formula: NaHCO₃
MW: 84.01 g/mol
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty