Would you like to log in again?
Sodium carbonate-bicarbonate eluent solution 3 mM Na₂CO₃/2 mM NaOH for ion chromatography
Supplier: Ricca Chemical
R2820000-1A R2820000-4A
RC28201EA 168.21 USD InStock
RC28201 RC282032
Sodium carbonate-bicarbonate eluent solution 3 mM Na₂CO₃/2 mM NaOH for ion chromatography
Sodium carbonate-bicarbonate eluent solution
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Certified Concentration 3 mM Na2CO3 - 2 mM NaOH
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty