Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eriochrome Black T USP Test Solution (TS)

Supplier: Ricca Chemical
Danger
Synonyms: 1-Naphthalenesulfonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1)1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1)Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonateEBTSolochrome Black TSodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulfonate

R2901000-120C R2901000-500C
RC2901-16EA 240.42 USD InStock
RC2901-16 RC2901-4
Eriochrome Black T USP Test Solution (TS)
Eriochrome Black T


Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
Boiling Pt: ~ 64.5 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003935
CAS Number: 1787-61-7
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, deep red liquid
Suitability for Use Wine Red (Excess Ca/Mg) - Blue (Excess EDTA)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty