Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eriochrome cyanine R USP Test Solution (TS)

Supplier: Ricca Chemical
Danger
Synonyms: Chromoxane Cyanine R

R2903000-500A R2903000-120A
RC2903-16EA 123.1 USD InStock
RC2903-16 RC2903-4
Eriochrome cyanine R USP Test Solution (TS)
Eriochrome cyanine R


Formula: C₂₃H₁₅Na₃O₉S
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00012411
CAS Number: 3564-18-9
Order Now
Specification Test Results
Appearance Clear, red, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty