Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fehling's reagent II (L(+)-potassium sodium tartrate in sodium hydroxide solution) for quantitative determination of reducing sugars

Supplier: Ricca Chemical
Danger
Synonyms: Fehling B solutionMonopotassium monosodium tartrate tetrahydrateRochelle Salt

R3000000-20F R3000000-4A R3000000-500A R3000000-1A
RC300016EA 63.91 USD InStock
RC300016 RC3000-1 RC3000-5 RC300032
Fehling's reagent II (L(+)-potassium sodium tartrate in sodium hydroxide solution) for quantitative determination of reducing sugars
Fehling's reagent II (L(+)-Potassium sodium tartrate in sodium hydroxide solution)


Density: 1.2 g/cm³ (20 °C)
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty