Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fehling's reagent I (copper(II) sulfate solution) for quantitative determination of reducing sugars

Supplier: Ricca Chemical
Synonyms: Cupric sulfate pentahydrateFehling A solutionCopper (II) sulfate 5-hydrate

R3010000-500A R3010000-4A R3010000-20F R3010000-1A
RC3010-5EA 407.69 USD InStock
RC3010-5 RC3010-1 RC301016 RC301032
Fehling's reagent I (copper(II) sulfate solution) for quantitative determination of reducing sugars
Fehling's reagent I (Copper (II) sulphate solution)


Formula: CuSO₄
MW: 159.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear, blue odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.2775 ± 0.0005 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty