Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ferric Ammonium Sulfate 8% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Warning
Synonyms: Ferric ammonium sulfate dodecahydrateAmmonium ferric sulfate dodecahydrateAmmonium iron sulfate dodecahydrateFerric ammonium sulphate dodecahydrateAmmonium iron sulphate dodecahydrateAmmonium ferric sulphate dodecahydrate

3058-32
RC3058-32EA 162.12 USD InStock
RC3058-32
Ferric Ammonium Sulfate 8% (w/v) in aqueous solution USP Test Solution (TS)
Ammonium iron (III) sulphate dodecahydrate


Formula: NH₄Fe(SO₄)₂·12H₂O
MW: 482.2 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150004
CAS Number: 7783-83-7
Order Now
Specification Test Results
Assay 8 ± 0.1 % (w/v)
NIST SRM 136

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty