Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium iron (III) sulfate saturated solution in water

Supplier: Ricca Chemical
Warning

R3070000-1A R3070000-500A R3070000-4A R3070000-120A
RC307032EA 225.33 USD InStock
RC307032 RC30704 RC307016 RC3070-1
Ammonium iron (III) sulfate saturated solution in water
Ammonium iron(III) sulphate solutions


Formula: NH₄Fe(SO₄)₂·12H₂O
MW: 482.2 g/mol
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150004
CAS Number: 7783-83-7
Order Now
Specification Test Results
Appearance Dark brown, odorless
Suitability for Use White precipitate (Excess Silver ions) - Reddish-Brown Color (Excess Thiocyante ions)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty