Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ferric chloride 9% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Danger
Synonyms: Iron(III) chloride solution

R3109000-500C R3109000-120C
RCR3109000120CEA 25.11 USD InStock
RCR3109000120C RC3109-16
Ferric chloride 9% (w/v) in aqueous solution USP Test Solution (TS)
Iron(III) chloride solutions


Formula: FeCl₃·6H₂O
MW: 270.3 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
UN: 2582
ADR: 8,III
Order Now
Specification Test Results
Appearance Bright orange/brown liquid
Assay (vs. Sodium Thiosulfate/Starch) 8.9-9.1 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty