Would you like to log in again?
Ferric chloride oxidizing solution 1% (w/v) aqueous, with 1.6% (w/v) sulfamic Acid for cyanide determination
Supplier: Ricca Chemical
Warning
R3113000-1A R3113000-500A
RC3113-32EA 82.17 USD InStock
RC3113-32 RC3113-16
Ferric chloride oxidizing solution 1% (w/v) aqueous, with 1.6% (w/v) sulfamic Acid for cyanide determination
Ferric chloride oxidizing solution
Order Now
Specification Test Results
Appearance Clear, yellowish-green, odorless
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty