Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(III) chloride 10% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger
Synonyms: Ferric chlorideIron trichloride

R3120000-120A R3120000-1A R3120000-500A
RC312016EA 72.37 USD InStock
RC312016 RC31204 RC312032
Iron(III) chloride 10% (w/v) in aqueous solution
Iron (III) chloride


Formula: FeCl₃
MW: 162.21 g/mol
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 2582
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, brown, no precipitate
Assay at 20 °C (traceable to NIST SRM 136) 10.00 ± 0.05 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty