Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ferric chloride ≥100% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger
Synonyms: Iron(III) chloride solution

R3130000-120A R3130000-500A
RC31304EA 119.58 USD InStock
RC31304 RC313016
Ferric chloride ≥100% (w/v) in aqueous solution
Iron(III) chloride solutions


Formula: FeCl₃·6H₂O
MW: 270.3 g/mol
Boiling Pt: ~ 105 °C
Density: 1.4700 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
UN: 2582
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, brown, odorless
Assay 100 ± 3 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty