Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(III) nitrate solution 404 g/L in dilute nitric acid

Supplier: Ricca Chemical
Danger

R3135000-1B R3135000-500B
RC3135-32EA 475.02 USD InStock
RC3135-32 RC3135-16
Iron(III) nitrate solution 404 g/L in dilute nitric acid
Iron(III) nitrate solutions


Formula: Fe(NO₃)₃·9H₂O
Boiling Pt: ~ 100 °C
Density: 1.2500 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149708
CAS Number: 7782-61-8
UN: 3264
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, pale yellow, slightly pungent odor
Assay at 20 °C (traceable to NIST SRM 136) 404.0 ± 0.4 g/L

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty