Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium iron (II) sulfate 22 g/L

Supplier: Ricca Chemical
Warning

R3151000-4C
RC3151-1EA 417.61 USD InStock
RC3151-1
Ammonium iron (II) sulfate 22 g/L
Haematoxylin Harris


Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0060 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
Order Now
Specification Test Results
Appearance Clear, light green, odorless
Assay at 20 °C (traceable to NIST SRM 136) 22.0 ± 0.5 g/L

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty