Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formaldehyde 8% (v/v) in aqueous solution histological fixative, (non-buffered)

Supplier: Ricca Chemical
Synonyms: MethylaldehydeFormolFormalinParaformaldehyde aqueous solution

R3178000-1A
RC3178-32EA 39.41 USD InStock
RC3178-32
Formaldehyde 8% (v/v) in aqueous solution histological fixative, (non-buffered)
Formaldehyde


Formula: CH₂O
MW: 30.03 g/mol
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 2209
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, colorless, Formaldehyde odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty