Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formaldehyde 10% (v/v) in aqueous solution histological fixative, (non-buffered)

Supplier: Ricca Chemical
Danger
Synonyms: MethylaldehydeFormolFormalinParaformaldehyde aqueous solution

R3180000-20F R3180000-10F R3180000-1A
RC318032EA 68.72 USD InStock
RC318032 RC31805 RC318025
Formaldehyde 10% (v/v) in aqueous solution histological fixative, (non-buffered)
Formaldehyde


Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 3334
ADR: 9,
Order Now
Specification Test Results
Appearance Clear, colorless, Formaldehyde odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty